Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,7-Dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate, 98%

$177.01 - $641.88

Chemical Identifiers

Chemical Name or Material 6,7-Dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate
CAS 4784-02-5
Molecular Formula C10H10N2O4·H2O
Formula Weight 240.22 (222.20 Anhydrous)
MDL Number MFCD09953682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2594103

 
alfa aesar™
H2594103
1g Each for $177.01

AAH2594106

 
alfa aesar™
H2594106
5g Each for $641.88
Description & Specifications

Specifications

Formula Weight 240.22 (222.20 Anhydrous)
Chemical Name or Material 6,7-Dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate
Assay 98%
Molecular Formula C10H10N2O4·H2O
Melting Point 352° to 354°C
MDL Number MFCD09953682
CAS 4784-02-5
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.