missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one, 98%

Manufacturer:  Alfa Aesar™ H3532103

 View more versions of this product

Catalog No. AAH3532103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one
1g
C10H8N2O
6,7-dihydrocyclopenta 4,5 pyrrolo 3,2-b pyridin-8 5h-one, 7,12-diazatricyclo 6.4.0.0 2 ,? dodeca-1 12 ,2 6 ,8,10-tetraen-3-one
C1CC(=O)C2=C1NC3=C2N=CC=C3
172.187
172.18
98%
Green
1421263-65-1
MFCD22988925
AMETXLSIVXCORJ-UHFFFAOYSA-N
6,7-dihydro-5H-cyclopenta[2,3]pyrrolo[2,4-b]pyridin-8-one
73995189
Solid

Chemical Identifiers

1421263-65-1
172.187
AMETXLSIVXCORJ-UHFFFAOYSA-N
73995189
C1CC(=O)C2=C1NC3=C2N=CC=C3
C10H8N2O
MFCD22988925
6,7-dihydrocyclopenta 4,5 pyrrolo 3,2-b pyridin-8 5h-one, 7,12-diazatricyclo 6.4.0.0 2 ,? dodeca-1 12 ,2 6 ,8,10-tetraen-3-one
6,7-dihydro-5H-cyclopenta[2,3]pyrrolo[2,4-b]pyridin-8-one
SDS Documents