Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one, 98%

Chemical Identifiers

Chemical Name or Material 6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one
Formula Weight 172.18
MDL Number MFCD22988925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3532103

  SDS
alfa aesar™
H3532103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH35321MD

  SDS
alfa aesar™
H35321MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.18
Chemical Name or Material 6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one
Physical Form Solid
Assay 98%
Color Green
MDL Number MFCD22988925
Quantity 1g

Applications
6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.