missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6,7-Dihydrobenzo[b]thiophen-4-one, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6022004
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
Solubility
Soluble in chloroform.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
6,7-Dihydrobenzo[b]thiophen-4-one | |
38°C to 40°C | |
110°C (230°F) | |
1.5895 | |
2 g | |
6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone | |
GJEKNELSXNSYAQ-UHFFFAOYSA-N | |
6,7-dihydro-5H-1-benzothiophen-4-one | |
83418 | |
98% |
13414-95-4 | |
120°C | |
C8H8OS | |
MFCD00005861 | |
112276 | |
Soluble in chloroform. | |
C1CC2=C(C=CS2)C(=O)C1 | |
152.211 | |
152.21 |
Chemical Identifiers
13414-95-4 | |
152.211 | |
GJEKNELSXNSYAQ-UHFFFAOYSA-N | |
83418 | |
C1CC2=C(C=CS2)C(=O)C1 |
C8H8OS | |
MFCD00005861 | |
6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone | |
6,7-dihydro-5H-1-benzothiophen-4-one |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 236-510-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only