Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,7-Dihydro-5-methylcyclopentapyrazine, 98%

$54.85 - $632.84

Chemical Identifiers

Chemical Name or Material 6,7-Dihydro-5-methylcyclopentapyrazine
CAS 23747-48-0
Molecular Formula C8H10N2
Formula Weight 134.18
MDL Number MFCD00040997
Synonym 5-Methyl-6,7-dihydrocyclopentapyrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2167103

  SDS
alfa aesar™
B2167103
1g Each for $54.85

AAB2167106

  SDS
alfa aesar™
B2167106
5g Each for $170.27

AAB2167114

  SDS
alfa aesar™
B2167114
25g Each for $632.84
Description & Specifications

Specifications

Formula Weight 134.18
Quantity 1g
Assay 98%
Synonym 5-Methyl-6,7-dihydrocyclopentapyrazine
TSCA TSCA
Health Hazard 1 Warning
Molecular Formula C8H10N2
MDL Number MFCD00040997
Chemical Name or Material 6,7-Dihydro-5-methylcyclopentapyrazine
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Boiling Point 201° to 203°C
CAS 23747-48-0
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.