Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione, 98%

Chemical Identifiers

Chemical Name or Material 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
CAS 67911-21-1
Formula Weight 140.14
MDL Number MFCD09751198
Beilstein 117391
Synonym Caronic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6689222

  SDS
alfa aesar™
H6689222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6689214

  SDS
alfa aesar™
H6689214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 53° to 57°C
Formula Weight 140.14
CAS 67911-21-1
Synonym Caronic anhydride
Chemical Name or Material 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
MDL Number MFCD09751198
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Moisture Sensitive
Physical Form Solid
Assay 98%
Color White
Health Hazard 1 Warning
Beilstein 117391
Boiling Point 137°C (14mmHg)
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.