missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6,6′-Dimethyl-2,2′-bipyridyl, TCI America™

$18.33 - $261.25

Chemical Identifiers

CAS 4411-80-7
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00059776
InChI Key OHJPGUSXUGHOGE-UHFFFAOYSA-N
Synonym 6,6′-Bi-2-picoline, 6,6′-Di-2-picolyl
PubChem CID 20445
IUPAC Name 2-methyl-6-(6-methylpyridin-2-yl)pyridine
SMILES CC1=CC=CC(=N1)C2=NC(=CC=C2)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B0842100MG View Documents TCI America
B0842100MG
100mg Each for $18.33
 
B08421G View Documents TCI America
B08421G
1g Each for $70.28
 
B08425G View Documents TCI America
B08425G
5g Each for $261.25
 
Specifications

Chemical Identifiers

4411-80-7
184.242
OHJPGUSXUGHOGE-UHFFFAOYSA-N
20445
CC1=CC=CC(=N1)C2=NC(=CC=C2)C
C12H12N2
MFCD00059776
6,6′-Bi-2-picoline, 6,6′-Di-2-picolyl
2-methyl-6-(6-methylpyridin-2-yl)pyridine

Specifications

286°C
88°C
4411-80-7
MFCD00059776
OHJPGUSXUGHOGE-UHFFFAOYSA-N
2-methyl-6-(6-methylpyridin-2-yl)pyridine
20445
Crystal-Powder at 20°C
6,6′-Dimethyl-2,2′-bipyridyl
White-Yellow
100mg
C12H12N2
6,6′C-Bi-2-picoline, 6,6′C-Di-2-picolyl
CC1=CC=CC(=N1)C2=NC(=CC=C2)C
184.242
184.24
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

RTECSNumber : DW1767000

TSCA : No