Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,6'-Dichloro-3,3'-bipyridine, 95%

$230.43 - $230.43

Chemical Identifiers

Chemical Name or Material 6,6'-Dichloro-3,3'-bipyridine
CAS 206438-08-6
Molecular Formula C10H6Cl2N2
Formula Weight 225.08g/mol
MDL Number MFCD09037430
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27064ME

 
alfa aesar™
H27064-ME
500mg Each for $230.43
Description & Specifications

Specifications

Quantity 500mg
Chemical Name or Material 6,6'-Dichloro-3,3'-bipyridine
CAS 206438-08-6
Assay 95%
Molecular Formula C10H6Cl2N2
Formula Weight 225.08g/mol
MDL Number MFCD09037430
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.