missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6,6'-Bis(chloromethyl)-2,2'-bipyridyl 98.0+%, TCI America™
Supplier: TCI America B46451G
Specifications
6,6′-Bis(chloromethyl)-2,2′-bipyridyl | |
162°C | |
C12H10Cl2N2 | |
1 g | |
6,6′C-Bis(chloromethyl)-2,2′C-bipyridine | |
ClCC1=CC=CC(=N1)C1=NC(CCl)=CC=C1 | |
253.13 | |
253.13 | |
Crystalline Powder |
74065-64-8 | |
White | |
MFCD02181082 | |
3261 | |
CUWSIBMCPYEBPL-UHFFFAOYSA-N | |
6,6'-bis(chloromethyl)-2,2'-bipyridine | |
1502060 | |
≥98.0% (GC,T) |
Chemical Identifiers
74065-64-8 | |
253.13 | |
CUWSIBMCPYEBPL-UHFFFAOYSA-N | |
1502060 | |
ClCC1=CC=CC(=N1)C1=NC(CCl)=CC=C1 |
C12H10Cl2N2 | |
MFCD02181082 | |
6,6′C-Bis(chloromethyl)-2,2′C-bipyridine | |
6,6'-bis(chloromethyl)-2,2'-bipyridine |
Safety and Handling
TSCA : No