missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene, TCI America™

$297.92 - $1,034.33

Chemical Identifiers

CAS 1429188-77-1
Molecular Formula C10H5F13
Molecular Weight (g/mol) 372.129
MDL Number MFCD28014057
InChI Key PUTFFSRJABOVAD-ONEGZZNKSA-N
PubChem CID 122360969
IUPAC Name (3E)-6,6,7,7,8,8,8-heptafluoro-5,5-bis(trifluoromethyl)octa-1,3-diene
SMILES C=CC=CC(C(C(C(F)(F)F)(F)F)(F)F)(C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H151525G View Documents TCI America
H151525G
25g Each for $1,034.33
 
H15155G View Documents TCI America
H15155G
5g Each for $297.92
 
Specifications

Chemical Identifiers

1429188-77-1
372.129
PUTFFSRJABOVAD-ONEGZZNKSA-N
(3E)-6,6,7,7,8,8,8-heptafluoro-5,5-bis(trifluoromethyl)octa-1,3-diene
C10H5F13
MFCD28014057
122360969
C=CC=CC(C(C(C(F)(F)F)(F)F)(F)F)(C(F)(F)F)C(F)(F)F

Specifications

Yellow
1429188-77-1
MFCD28014057
PUTFFSRJABOVAD-ONEGZZNKSA-N
(3E)-6,6,7,7,8,8,8-heptafluoro-5,5-bis(trifluoromethyl)octa-1,3-diene
122360969
Clear Liquid at 20°C
6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene
25g
C10H5F13
1993
C=CC=CC(C(C(C(F)(F)F)(F)F)(F)F)(C(F)(F)F)C(F)(F)F
372.129
372.13
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator