Please login to your online account to display your discounted pricing

(6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedionato)silver, ≥99%, ACROS Organics™

$67.09 - $67.09

Chemical Identifiers

Chemical Name or Material (6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedionato)silver
CAS 76121-99-8
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H10AgF7O2
Linear Formula CF3CF2CF2COCH=CH(OAg)C(CH3)3
Formula Weight 403.04
MDL Number MFCD00010348
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC208690010

  SDS
thermo fisher scientific
208690010
Glass bottle 1g Each for $67.09
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 403.04
Physical Form Fine Crystalline Powder
Quantity 1g
Color Green-brown to brown
CAS Min % 99.0
CAS Max % 100.0
Molecular Formula C10H10AgF7O2
Infrared Spectrum Authentic
Assay Percent Range 99+%
CAS 76121-99-8
Chemical Name or Material (6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedionato)silver
MDL Number MFCD00010348
Synonym Ag(fod)
Linear Formula CF3CF2CF2COCH=CH(OAg)C(CH3)3
Packaging Glass bottle