missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Simport™ Scientific 6.5mL HDPE Snaptwist™ Scintillation Vial

With polypropylene cap.Simport™Scientific 6.5mL HDPE Snaptwist™ Scintillation Vials are designed for use with liquid scintillation and gamma counting, chromatography, sample storage and culturing.

Manufacturer:  Simport™ Scientific S207

Catalog No. 03-342-3



Description

Description

  • Shoulderless vial features full-width 12.5mm opening
  • Snaptwist closure can be easily removed with a 1/4 turn
Specifications

Specifications

0.62 in.
HDPE
Used for liquid scintillation counting, gamma counting, chromatography, sample storage and culturing, fit into the LKB and packard Varisette counters
0.21 oz.
57 mm
16 mm
6.5 mL
2.24 in.
Polypropylene
SDS
Documents