Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(4-Morpholinylamino)pyridine-3-boronic acid pinacol ester, 95%

$396.32 - $396.32

Chemical Identifiers

Chemical Name or Material 6-(4-Morpholinylamino)pyridine-3-boronic acid pinacol ester
CAS 1073354-38-7
Molecular Formula C15H24BN3O3
Formula Weight 305.19g/mol
MDL Number MFCD06798279
Synonym N-(4-Morpholinyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2697103

 
alfa aesar™
H26971-03
1g Each for $396.32
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 6-(4-Morpholinylamino)pyridine-3-boronic acid pinacol ester
CAS 1073354-38-7
Assay 95%
Molecular Formula C15H24BN3O3
Formula Weight 305.19g/mol
MDL Number MFCD06798279
Synonym N-(4-Morpholinyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.