missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester

$97.59 - $221.66

Chemical Identifiers

CAS 485799-04-0
Molecular Formula C15H23BN2O3
Formula Weight 290.17
MDL Number MFCD04039874
Synonym 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine; 6-(4-Morpholino)pyridine-3-boronic acid pinacol ester
Chemical Name or Material 6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5014303 SDS Alfa Aesar™
H5014303
1g Each for $221.66
AAH50143MD SDS Alfa Aesar™
H50143MD
250mg Each for $97.59
Specifications Description & Specifications

Chemical Identifiers

485799-04-0
290.17
4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine; 6-(4-Morpholino)pyridine-3-boronic acid pinacol ester
C15H23BN2O3
MFCD04039874
6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester

Specifications

485799-04-0
290.17
4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine; 6-(4-Morpholino)pyridine-3-boronic acid pinacol ester
6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester
C15H23BN2O3
MFCD04039874
1g

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning