Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(4-Fluorophenyl)pyridine-3-boronic acid pinacol ester, 95%

$268.20 - $620.78

Chemical Identifiers

Chemical Name or Material 6-(4-Fluorophenyl)pyridine-3-boronic acid pinacol ester
CAS 1073354-81-0
Molecular Formula C17H19BFNO2
Formula Weight 299.15
MDL Number MFCD09037494
Synonym 2-(4-Fluorophenyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2716803

  SDS
alfa aesar™
H2716803
1g Each for $620.78

AAH27168MD

  SDS
alfa aesar™
H27168MD
250mg Each for $268.20
Description & Specifications

Specifications

Formula Weight 299.15
Synonym 2-(4-Fluorophenyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C17H19BFNO2
MDL Number MFCD09037494
CAS 1073354-81-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 6-(4-Fluorophenyl)pyridine-3-boronic acid pinacol ester
Assay 95%
Health Hazard 1 Warning
Quantity 1g

Applications
Used in organic synthesis. The boronic esters are widely used for carrying out cross coupling reactions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.