Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(4-Ethoxycarbonyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester, 95%

$230.54 - $521.34

Chemical Identifiers

Chemical Name or Material 6-(4-Ethoxycarbonyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester
CAS 1073354-26-3
Molecular Formula C18H28BN3O4
Formula Weight 361.25
MDL Number MFCD06798269
Synonym 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]piperazine-1-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2712203

  SDS
alfa aesar™
H2712203
1g Each for $521.34

AAH27122MD

  SDS
alfa aesar™
H27122MD
250mg Each for $230.54
Description & Specifications

Specifications

Formula Weight 361.25
Quantity 1g
MDL Number MFCD06798269
Assay 95%
Synonym 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]piperazine-1-carboxylic acid ethyl ester
CAS 1073354-26-3
Molecular Formula C18H28BN3O4
Chemical Name or Material 6-(4-Ethoxycarbonyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester
Melting Point 135° to 139°C

Applications
6-(4-Ethoxycarbonyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.