Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(4-Bromophenyl)-4,5-dihydro-3(2H)-pyridazinone, 98%

$38.62 - $150.85

Chemical Identifiers

Chemical Name or Material 6-(4-Bromophenyl)-4,5-dihydro-3(2H)-pyridazinone
CAS 36725-37-8
Molecular Formula C10H9BrN2O
Formula Weight 253.1
MDL Number MFCD00203490
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5058803

  SDS
alfa aesar™
H5058803
1g Each for $38.62

AAH5058806

  SDS
alfa aesar™
H5058806
5g Each for $150.85
Description & Specifications

Specifications

Formula Weight 253.1
Health Hazard 1 Warning
Chemical Name or Material 6-(4-Bromophenyl)-4,5-dihydro-3(2H)-pyridazinone
Assay 98%
Molecular Formula C10H9BrN2O
MDL Number MFCD00203490
CAS 36725-37-8
Melting Point 167° to 171°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.