Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(4-Boc-1-piperazinyl)pyridine-3-boronic acid pinacol ester, 99%

$254.65 - $254.65

Chemical Identifiers

Chemical Name or Material 6-(4-Boc-1-piperazinyl)pyridine-3-boronic acid pinacol ester
CAS 496786-98-2
Molecular Formula C20H32BN3O4
Formula Weight 389.31
MDL Number MFCD04039875
Synonym tert-Butyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]piperazine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5014503

  SDS
alfa aesar™
H5014503
1g Each for $254.65
Description & Specifications

Specifications

Formula Weight 389.31
Synonym tert-Butyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]piperazine-1-carboxylate
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
MDL Number MFCD04039875
CAS 496786-98-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Molecular Formula C20H32BN3O4
Chemical Name or Material 6-(4-Boc-1-piperazinyl)pyridine-3-boronic acid pinacol ester
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.