Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(4-Benzyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester, 95%

$189.34 - $534.14

Chemical Identifiers

Chemical Name or Material 6-(4-Benzyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester
CAS 1015242-03-1
Molecular Formula C22H30BN3O2
Formula Weight 379.31
MDL Number MFCD06798275
Synonym 1-Benzyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27794MD

  SDS
alfa aesar™
H27794MD
250mg Each for $189.34

AAH2779403

  SDS
alfa aesar™
H2779403
1g Each for $534.14
Description & Specifications

Specifications

Formula Weight 379.31
Chemical Name or Material 6-(4-Benzyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester
CAS 1015242-03-1
Assay 95%
Molecular Formula C22H30BN3O2
MDL Number MFCD06798275
Synonym 1-Benzyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]piperazine
Melting Point 128° to 131°C
Health Hazard 1 Warning
Quantity 250mg
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.