missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444970010
Specifications
6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester | |
34°C to 37°C | |
918643-56-8 | |
C16H27BN2O3 | |
QWJUJXPYTLOKRL-UHFFFAOYSA-N | |
N,N-dimethyl-3-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxypropan-1-amine | |
17998906 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
2-3-n,n-dimethylamino-propoxy pyridine-5-boronic acid, pinacol ester, dimethy 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-22-yloxy propyl amine, dimethyl 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propyl amine, dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yloxy-propyl-amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propan-1-amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yloxy propan-1-amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl oxy-1-propanamine | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCCCN(C)C | |
306.21 | |
306.21 |
Chemical Identifiers
918643-56-8 | |
306.21 | |
2-3-n,n-dimethylamino-propoxy pyridine-5-boronic acid, pinacol ester, dimethy 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-22-yloxy propyl amine, dimethyl 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propyl amine, dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yloxy-propyl-amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propan-1-amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yloxy propan-1-amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl oxy-1-propanamine | |
N,N-dimethyl-3-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxypropan-1-amine |
C16H27BN2O3 | |
QWJUJXPYTLOKRL-UHFFFAOYSA-N | |
17998906 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCCCN(C)C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning