missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[6-(2-Furyl)pyrid-3-yl]methylamine, 90%, Maybridge

Chemical Identifiers

CAS 441055-75-0
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C10H10N2O
Formula Weight 174.2
MDL Number MFCD09817484
Chemical Name or Material [6-(2-Furyl)pyrid-3-yl]methylamine
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC54913CB SDS Fisher Scientific Company
CC54913CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC54913DA SDS Fisher Scientific Company
CC54913DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

441055-75-0
100.0
C10H10N2O
MFCD09817484
90%
89.5
90%
174.2
[6-(2-Furyl)pyrid-3-yl]methylamine

Specifications

441055-75-0
100.0
Amber Glass Bottle
174.2
250mg
90%
89.5
90%
C10H10N2O
MFCD09817484
[6-(2-Furyl)pyrid-3-yl]methylamine

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger