missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(2-furyl)nicotinic acid, Maybridge

Manufacturer:  Fisher Scientific Company CC54901DE

 View more versions of this product

Catalog No. CC54901DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

189.17
97%
6-(2-Furyl)nicotinic acid
97%
5g
Amber glass bottle
884507-36-2
C10H7NO3

Chemical Identifiers

97%
189.17
97%
C10H7NO3
6-(2-Furyl)nicotinic acid

Safety and Handling

GHS Signal Word: Warning