Please login to your online account to display your discounted pricing

6-(2-furyl)nicotinic acid, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC54901CB

 View more versions of this product

Catalog No. CC54901CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 189.17
Quantity 250mg
Assay 97%
Chemical Name or Material 6-(2-Furyl)nicotinic acid
Assay Percent Range 97%
Molecular Formula C10H7NO3
CAS 884507-36-2
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 6-(2-Furyl)nicotinic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7NO3
Formula Weight 189.17
Accessories

GHS Signal Word: Warning