Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,2'-Dichloro-3,4'-bipyridine, 95%

$268.49 - $268.49

Chemical Identifiers

Chemical Name or Material 6,2'-Dichloro-3,4'-bipyridine
CAS 942206-22-6
Molecular Formula C10H6Cl2N2
Formula Weight 225.08g/mol
MDL Number MFCD09037432
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27379ME

 
alfa aesar™
H27379-ME
500mg Each for $268.49
Description & Specifications

Specifications

Melting Point 221° to 223°C
Quantity 500mg
Chemical Name or Material 6,2'-Dichloro-3,4'-bipyridine
CAS 942206-22-6
Assay 95%
Molecular Formula C10H6Cl2N2
Formula Weight 225.08g/mol
MDL Number MFCD09037432
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.