missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6,2',4'-Trimethoxyflavone, Tocris™

Aryl hydrocarbon receptor antagonist

$142.08 - $598.33

Chemical Identifiers

Assay Percent Range >97%
Molecular Formula C18H16O5
Formula Weight 312.32
Synonym 2', 4', 6-Trimethoxyflavone
Chemical Name or Material 6, 2', 4'-Trimethoxyflavone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
385950 R & D Systems
RDS385950
50mg Each for $598.33
385910 R & D Systems
RDS385910
10mg Each for $142.08
Specifications Description & Specifications

Chemical Identifiers

>97%
312.32
6, 2', 4'-Trimethoxyflavone
C18H16O5
2', 4', 6-Trimethoxyflavone

Specifications

50mg
>97%
312.32
6, 2', 4'-Trimethoxyflavone
720675-74-1
C18H16O5
2', 4', 6-Trimethoxyflavone

Safety and Handling

Recommended Storage : Store at Room Temperature