Please login to your online account to display your discounted pricing

6,13-Pentacenequinone, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC364270050

 View more versions of this product

Catalog No. AC364270050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Powder
Formula Weight 308.34
Quantity 5g
Color Yellow
Beilstein 07, IV, 2689
CAS Max % 100.0
Chemical Name or Material 6, 13-Pentacenequinone
Name Note 99%
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C22H12O2
MDL Number MFCD00003709
Packaging Glass bottle
CAS 3029-32-1
Synonym 6, 13-Pentacenedione
CAS Min % 98.5

Specifications

Chemical Name or Material 6, 13-Pentacenequinone
Name Note 99%
CAS 3029-32-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C22H12O2
Formula Weight 308.34
MDL Number MFCD00003709
Synonym 6, 13-Pentacenedione