missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6,13-Pentacenequinone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 364270010

 View more versions of this product

Catalog No. AC364270010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6, 13-Pentacenequinone
3029-32-1
100.0
Yellow
98.5% min. (HPLC)
C22H12O2
1 g
6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa
UFCVADNIXDUEFZ-UHFFFAOYSA-N
pentacene-6,13-dione
76415
99%
99%
98.5
394.0°C
Authentic
Glass bottle
MFCD00003709
07,I,451
Solubility in water: insoluble. Other solubilities: soluble in ethanol,benzene,pyridine,dioxane,,dimethylformamide,nitrobenzene
C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O
308.34
308.34
Powder

Chemical Identifiers

3029-32-1
308.34
UFCVADNIXDUEFZ-UHFFFAOYSA-N
76415
C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O
C22H12O2
MFCD00003709
6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa
pentacene-6,13-dione
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only