Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(1-Pyrrolidinyl)pyridine-3-carboxaldehyde, 98%

$244.74 - $1,086.73

Chemical Identifiers

Chemical Name or Material 6-(1-Pyrrolidinyl)pyridine-3-carboxaldehyde
CAS 261715-39-3
Molecular Formula C10H12N2O
Formula Weight 176.22
MDL Number MFCD06200881
Synonym 6-(1-Pyrrolidino)nicotinaldehyde; 6-(1-Pyrrolidino)pyridine-3-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5012603

 
alfa aesar™
H5012603
1g Each for $244.74

AAH5012606

 
alfa aesar™
H5012606
5g Each for $1,086.73
Description & Specifications

Specifications

Formula Weight 176.22
Chemical Name or Material 6-(1-Pyrrolidinyl)pyridine-3-carboxaldehyde
Assay 98%
Molecular Formula C10H12N2O
MDL Number MFCD06200881
CAS 261715-39-3
Synonym 6-(1-Pyrrolidino)nicotinaldehyde; 6-(1-Pyrrolidino)pyridine-3-carboxaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.