Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(1-Piperidinyl)pyridine-3-carboxaldehyde, 98%

$289.30 - $1,288.28

Chemical Identifiers

Chemical Name or Material 6-(1-Piperidinyl)pyridine-3-carboxaldehyde
CAS 241816-11-5
Molecular Formula C11H14N2O
Formula Weight 190.25
MDL Number MFCD04117814
Synonym 6-(1-Piperidino)pyridine-3-carboxaldehyde; 6-(1-Piperidinyl)nicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5012706

  SDS
alfa aesar™
H5012706
5g Each for $1,288.28

AAH5012703

  SDS
alfa aesar™
H5012703
1g Each for $289.30
Description & Specifications

Specifications

Formula Weight 190.25
Quantity 5g
MDL Number MFCD04117814
Assay 98%
Chemical Name or Material 6-(1-Piperidinyl)pyridine-3-carboxaldehyde
Synonym 6-(1-Piperidino)pyridine-3-carboxaldehyde; 6-(1-Piperidinyl)nicotinaldehyde
Melting Point 42° to 44°C
CAS 241816-11-5
Molecular Formula C11H14N2O
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.