Please login to your online account to display your discounted pricing

Alfa Aesar™ (5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone, 98%

Chemical Identifiers

Chemical Name or Material (5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone
CAS 100516-54-9
Formula Weight 387.43
MDL Number MFCD00074958
Synonym (2R,3S)-(-)-N-Benzyloxycarbonyl-6-oxo-2,3-diphenylmorpholine; Benzyl (2R,3S)-(-)-6-oxo-2,3-diphenylmorpholine-4-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6082103

  SDS
alfa aesar™
H6082103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 205° to 207°C
Formula Weight 387.43
Synonym (2R,3S)-(-)-N-Benzyloxycarbonyl-6-oxo-2,3-diphenylmorpholine; Benzyl (2R,3S)-(-)-6-oxo-2,3-diphenylmorpholine-4-carboxylate
MDL Number MFCD00074958
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
CAS 100516-54-9
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
Chemical Name or Material (5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone
Physical Form Solid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C