Please login to your online account to display your discounted pricing

Alfa Aesar™ 5H-Perfluoropentanoic acid, 96%

Chemical Identifiers

Chemical Name or Material 5H-Perfluoropentanoic acid
CAS 376-72-7
Molecular Formula H(CF2)4CO2H
Formula Weight 246.06
MDL Number MFCD00054690
UN Number UN3265
Beilstein 1797233
Synonym 5H-Octafluoropentanoic acid; 5H-Octafluorovaleric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1696006

  SDS
alfa aesar™
L1696006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1696014

  SDS
alfa aesar™
L1696014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.06
Chemical Name or Material 5H-Perfluoropentanoic acid
CAS 376-72-7
MDL Number MFCD00054690
Assay 96%
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Molecular Formula H(CF2)4CO2H
Quantity 5g
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 0°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Beilstein 1797233
Boiling Point 165° to 166°C
RTECS Number YV7799000
UN Number UN3265
Synonym 5H-Octafluoropentanoic acid; 5H-Octafluorovaleric acid
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.