Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
$424.27 - $481.24
Specifications
MWCO | 3.5 kDa |
---|
Includes:
- 1m or 5m of Spectra/Por 3 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity | |||
08-700-163
|
Spectrum™
132724T |
6.4 mL/cm vol./length | 29 mm | 5 m Tubing/Kit |
Each for $424.27
|
|
|||
08-801-245
|
Spectrum™
132720T |
1.1 mL/cm vol./length | 11.5 mm | 5 m Tubing/Kit |
Each for $428.00
|
|
|||
08-801-246
|
Spectrum™
132725T |
5 m | 34 mm | 5 m Tubing/Kit |
Each for $481.24
|
|
|||
Description
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Specifications
3.5 kDa |