missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Spectrum™ Labs 50 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
$428.94 - $496.55
Specifications
MWCO | 50 kDa |
---|
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
08-700-128
|
Spectrum™
131381T |
1.8 mL/cm vol./length | 15 mm |
Each for $470.91
|
|
||||
08-700-127
|
Spectrum™
131372T |
0.32 mL/cm vol./length | 6.4 mm |
Each for $428.94
|
|
||||
08-700-129
|
Spectrum™
131384T |
3.1 mL/cm vol./length | 20 mm |
Each for $480.24
|
|
||||
08-801-252
|
Spectrum™
131378T |
0.79 mL/cm vol./length | 10 mm |
Each for $496.55
|
|
||||
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
50 kDa |