missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ iBind™ Flex Western Starter Kit
Allows better antigen-antibody interaction and solutions flow between the iBind™ Card and membrane for maximum interaction between protein and antibody
Supplier: Invitrogen™ SLF2000S
Description
The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.The Invitrogen™ iBind™ Flex Western System offers:
- Flexible options—optimize antibody use, change formats and reduce hands-on time
- Cost savings—use up to 80% less primary antibody
- Reproducibility—automated processing provides improved blot-to-blot consistency
Specifications
• 1 iBind Flex Western Device • 10 iBind Flex Cards, store at room temperature. • 1 iBind Flex Solution Kit, store at 2–8°C. |
|
one midi blot, six vertically-cut strip blots, two mini blots | |
Nitrocellulose, PVDF | |
iBind™ | |
Starter Kit |
∼2.5 hours | |
Room Temperature | |
Chromogenic, Chemiluminescent, Fluorescent | |
10 iBind cards, 1 iBind solution kit, Western device |
For Research Use Only. Not for use in diagnostic procedures.