missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Vinyluracil, 97%

Manufacturer:  Alfa Aesar™ 4437901

 View more versions of this product

Catalog No. AA4437901



Specifications

Specifications

5-Vinyluracil
37107-81-6
MFCD00913267
ZRYZBEQILKESAW-UHFFFAOYSA-N
5-ethenyl-1H-pyrimidine-2,4-dione
99305
Powder
0.25g
C6H6N2O2
2,4 1h,3h-pyrimidinedione,5-ethenyl
C=CC1=CNC(=O)NC1=O
138.126
138.12
97%

Chemical Identifiers

37107-81-6
138.126
ZRYZBEQILKESAW-UHFFFAOYSA-N
99305
C=CC1=CNC(=O)NC1=O
C6H6N2O2
MFCD00913267
2,4 1h,3h-pyrimidinedione,5-ethenyl
5-ethenyl-1H-pyrimidine-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents