missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Vinyl-2-norbornene, 98%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 164865000

 View more versions of this product

Catalog No. AC164865000



Specifications

Specifications

5-Vinyl-2-norbornene
Authentic
1.4800 to 1.482
141°C
-80°C
98%
97.5
97.5% min. (GC)
MFCD00080691
INYHZQLKOKTDAI-UHFFFAOYSA-N
5-ethenylbicyclo[2.2.1]hept-2-ene
18273
Liquid
27°C
Glass bottle
100 to 200ppm t-butylcatechol
Undesignated
500mL
3048-64-4
100.0
C9H12
2-norbornene, 5-vinyl
C=CC1CC2CC1C=C2
120.19
120.19
98%

Chemical Identifiers

3048-64-4
120.19
INYHZQLKOKTDAI-UHFFFAOYSA-N
18273
C=CC1CC2CC1C=C2
C9H12
MFCD00080691
2-norbornene, 5-vinyl
5-ethenylbicyclo[2.2.1]hept-2-ene
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 221-259-8

RTECSNumber : RC0350000

TSCA : TSCA

SDS Documents