Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Undecanone, 99%

$320.95 - $320.95

Chemical Identifiers

Chemical Name or Material 5-Undecanone
CAS 33083-83-9
Molecular Formula C11H22O
Formula Weight 170.3
MDL Number MFCD00048899
Beilstein 1749497
Synonym n-Butyl n-hexyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2033614

  SDS
alfa aesar™
B2033614
25g Each for $320.95
Description & Specifications

Specifications

Formula Weight 170.3
MDL Number MFCD00048899
Synonym n-Butyl n-hexyl ketone
Chemical Name or Material 5-Undecanone
Molecular Formula C11H22O
CAS 33083-83-9
Beilstein 1749497
Quantity 25g
Melting Point 1° to 2°C
Boiling Point 103° to 106°C (12mmHg)
Assay 99%