Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Undecanol, 98%

$28.64 - $118.58

Chemical Identifiers

Chemical Name or Material 5-Undecanol
CAS 37493-70-2
Molecular Formula C11H24O
Formula Weight 172.31
MDL Number MFCD00046719
Beilstein 1698382
Synonym n-Butyl n-hexyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0401903

  SDS
alfa aesar™
L0401903
1g Each for $28.64

AAL0401906

  SDS
alfa aesar™
L0401906
5g Each for $118.58
Description & Specifications

Specifications

Formula Weight 172.31
Chemical Name or Material 5-Undecanol
CAS 37493-70-2
MDL Number MFCD00046719
Molecular Formula C11H24O
Beilstein 1698382
Melting Point −4°C
Quantity 1g
Assay 98%
Synonym n-Butyl n-hexyl carbinol
Boiling Point 139° to 140°C (17mmHg)