missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Trimethylsilyl-4-pentyn-1-ol, 97%, ACROS Organics™

Chemical Identifiers

CAS 13224-84-5
Molecular Formula C8H16OSi
Molecular Weight (g/mol) 156.3
MDL Number MFCD00190229
InChI Key LIHAMNSEAAPIRM-UHFFFAOYSA-N
Synonym 4-pentyn-1-ol, 5-trimethylsilyl, 4-pentyn-1-ol,5-trimethylsilyl, 5-trimethylsilyl ;pent-4-yn-1-ol, 5-trimethylsilyl pent-4-yn-1-ol, 5-trimethylsilyl-4-pentyn-1-ol, 5-trimethylsilyl-4-prntyn-1-ol, trimethyl 5-hydroxy-1-pentynyl silane
PubChem CID 2760861
IUPAC Name 5-trimethylsilylpent-4-yn-1-ol
SMILES C[Si](C)(C)C#CCCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC381330050
View Documents
Thermo Fisher Scientific
AC381330050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13224-84-5
156.3
LIHAMNSEAAPIRM-UHFFFAOYSA-N
2760861
C[Si](C)(C)C#CCCCO
C8H16OSi
MFCD00190229
4-pentyn-1-ol, 5-trimethylsilyl, 4-pentyn-1-ol,5-trimethylsilyl, 5-trimethylsilyl ;pent-4-yn-1-ol, 5-trimethylsilyl pent-4-yn-1-ol, 5-trimethylsilyl-4-pentyn-1-ol, 5-trimethylsilyl-4-prntyn-1-ol, trimethyl 5-hydroxy-1-pentynyl silane
5-trimethylsilylpent-4-yn-1-ol

Specifications

0.875g/mL
Glass bottle
Colorless to Yellow
13224-84-5
100.0
C8H16OSi
MFCD00190229
LIHAMNSEAAPIRM-UHFFFAOYSA-N
5-trimethylsilylpent-4-yn-1-ol
2760861
Liquid
5-Trimethylsilyl-4-pentyn-1-ol
Authentic
1.4560 to 1.4580 (20°C, 589nm)
5g
96.0
97%
(CH3)3SiC2C(CH2)3OH
4-pentyn-1-ol, 5-trimethylsilyl, 4-pentyn-1-ol,5-trimethylsilyl, 5-trimethylsilyl ;pent-4-yn-1-ol, 5-trimethylsilyl pent-4-yn-1-ol, 5-trimethylsilyl-4-pentyn-1-ol, 5-trimethylsilyl-4-prntyn-1-ol, trimethyl 5-hydroxy-1-pentynyl silane
C[Si](C)(C)C#CCCCO
156.3
156.3
≥96.0%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS