missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Trimethylsilyl-4-pentyn-1-ol, 97%

Manufacturer:  Alfa Aesar™ H5345706

 View more versions of this product

Catalog No. AAH5345706



Specifications

Specifications

5-Trimethylsilyl-4-pentyn-1-ol
C8H16OSi
MFCD00190229
LIHAMNSEAAPIRM-UHFFFAOYSA-N
5-trimethylsilylpent-4-yn-1-ol
2760861
97%
1.457
5g
13224-84-5
HO(CH2)3C≡CSi(CH3)3
4-pentyn-1-ol, 5-trimethylsilyl
C[Si](C)(C)C#CCCCO
156.3
156.3
0.861
76°C (15mmHg)
84°C (183°F)

Chemical Identifiers

13224-84-5
156.3
LIHAMNSEAAPIRM-UHFFFAOYSA-N
2760861
C[Si](C)(C)C#CCCCO
C8H16OSi
MFCD00190229
4-pentyn-1-ol, 5-trimethylsilyl
5-trimethylsilylpent-4-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents