missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Trifluoromethyl)uracil, TCI America™

Manufacturer:  TCI America T16225G

 View more versions of this product

Catalog No. T16225G



Specifications

Specifications

5-(Trifluoromethyl)uracil
5g
C5H3F3N2O2
5-trifluoromethyl uracil, 5-trifluoromethyluracil, 5-trifluoromethyl pyrimidine-2,4 1h,3h-dione, trifluoromethyluracil, 2,4 1h,3h-pyrimidinedione, 5-trifluoromethyl, 5-tfmu, uracil, 5-trifluoromethyl, 5-trifluoromethyl pyrimidine-2,4-diol, 5-trifluoromethyl uracil, 5-trifluoromethyl-uracil
C1=C(C(=O)NC(=O)N1)C(F)(F)F
180.086
180.09
≥97.0% (HPLC)
White-Yellow
54-20-6
MFCD00006024
LMNPKIOZMGYQIU-UHFFFAOYSA-N
5-(trifluoromethyl)-1H-pyrimidine-2,4-dione
5899
Crystal-Powder at 20°C

Chemical Identifiers

54-20-6
180.086
LMNPKIOZMGYQIU-UHFFFAOYSA-N
5899
C1=C(C(=O)NC(=O)N1)C(F)(F)F
C5H3F3N2O2
MFCD00006024
5-trifluoromethyl uracil, 5-trifluoromethyluracil, 5-trifluoromethyl pyrimidine-2,4 1h,3h-dione, trifluoromethyluracil, 2,4 1h,3h-pyrimidinedione, 5-trifluoromethyl, 5-tfmu, uracil, 5-trifluoromethyl, 5-trifluoromethyl pyrimidine-2,4-diol, 5-trifluoromethyl uracil, 5-trifluoromethyl-uracil
5-(trifluoromethyl)-1H-pyrimidine-2,4-dione
Safety and Handling

Safety and Handling

RTECSNumber : YR1750000

TSCA : No

Documents