Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Trifluoromethyl)uracil, 97%

$65.73 - $65.73

Chemical Identifiers

Chemical Name or Material 5-(Trifluoromethyl)uracil
CAS 54-20-6
Molecular Formula C5H3F3N2O2
Formula Weight 180.08
MDL Number MFCD00006024
Beilstein 612694
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL16196MD

  SDS
alfa aesar™
L16196MD
250mg Each for $65.73
Description & Specifications

Specifications

Formula Weight 180.08
Melting Point 251° to 254°C
Chemical Name or Material 5-(Trifluoromethyl)uracil
Assay 97%
Molecular Formula C5H3F3N2O2
MDL Number MFCD00006024
CAS 54-20-6
RTECS Number YR1970000
Beilstein 612694
Quantity 250mg