missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%

$45.95 - $746.39

Chemical Identifiers

CAS 80194-69-0
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD04113632
InChI Key NJHGVAYLDHROPT-UHFFFAOYSA-N
Synonym 5-trifluoromethyl pyridine-2-carboxylic acid, 5-trifluoromethyl picolinic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl pyridine-2-carboxylicacid, pubchem15164, acmc-209pj2
PubChem CID 10910349
IUPAC Name 5-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1C(F)(F)F)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6493703 View Documents Alfa Aesar™
H6493703
1g Each for $45.95
 
AAH6493706 View Documents Alfa Aesar™
H6493706
5g Each for $180.43
 
AAH6493714 View Documents Alfa Aesar™
H6493714
25g Each for $746.39
 
Description

Description

Applications
5-(Trifluoromethyl)pyridine-2-carboxylic acid is an intermediate used in the synthesis of β-secretase (BACE) inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

80194-69-0
191.109
NJHGVAYLDHROPT-UHFFFAOYSA-N
10910349
C1=CC(=NC=C1C(F)(F)F)C(=O)O
C7H4F3NO2
MFCD04113632
5-trifluoromethyl pyridine-2-carboxylic acid, 5-trifluoromethyl picolinic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl pyridine-2-carboxylicacid, pubchem15164, acmc-209pj2
5-(trifluoromethyl)pyridine-2-carboxylic acid

Specifications

Brown
1g
C7H4F3NO2
5-trifluoromethyl pyridine-2-carboxylic acid, 5-trifluoromethyl picolinic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl pyridine-2-carboxylicacid, pubchem15164, acmc-209pj2
C1=CC(=NC=C1C(F)(F)F)C(=O)O
191.109
191.11
95%
133°C to 137°C
80194-69-0
MFCD04113632
NJHGVAYLDHROPT-UHFFFAOYSA-N
5-(trifluoromethyl)pyridine-2-carboxylic acid
10910349
Powder
5-(Trifluoromethyl)pyridine-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning