Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Trifluoromethyl-m-phenylenediamine, 97+%

Chemical Identifiers

Chemical Name or Material 5-Trifluoromethyl-m-phenylenediamine
CAS 368-53-6
Molecular Formula C7H7F3N2
Formula Weight 176.14
MDL Number MFCD00014776
UN Number UN2811
Beilstein 2966748
Synonym 3,5-Diaminobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2366803

  SDS
alfa aesar™
B2366803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2366806

  SDS
alfa aesar™
B2366806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.14
Chemical Name or Material 5-Trifluoromethyl-m-phenylenediamine
Assay ≥97%
Molecular Formula C7H7F3N2
MDL Number MFCD00014776
CAS 368-53-6
UN Number UN2811
Beilstein 2966748
Synonym 3,5-Diaminobenzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H318-H302-H332-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number CZ1606000
Melting Point 86° to 90°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H318-H302-H332-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.