Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Trifluoromethyl)indole, 98%

$168.76 - $516.82

Chemical Identifiers

Chemical Name or Material 5-(Trifluoromethyl)indole
CAS 100846-24-0
Molecular Formula C9H6F3N
Formula Weight 185.15
MDL Number MFCD03095341
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH28782ME

  SDS
alfa aesar™
H28782ME
500mg Each for $516.82

AAH28782MC

  SDS
alfa aesar™
H28782MC
100mg Each for $168.76
Description & Specifications

Specifications

Formula Weight 185.15
Health Hazard 1 Warning
Chemical Name or Material 5-(Trifluoromethyl)indole
Assay 98%
Quantity 500mg
Melting Point 68° to 71°C
CAS 100846-24-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H6F3N
MDL Number MFCD03095341

Applications
In 3-Naphthylindole construction by rhodium (II)-catalyzed regioselective direct arylation of indoles with 1-diazonaphthalen-2-(1H)-ones treatment of 5-(trifluoromethyl)indole(1k) or 6-(trifluoromethyl)indole (1l) with 2a afforded the desired products 3k (86%)and 3l (86%).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.