Please login to your online account to display your discounted pricing

[5-(Trifluoromethyl)-2-furyl]methanol, ≥97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC49809DA

 View more versions of this product

Catalog No. CC49809DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 166.1
Quantity 1g
Assay 97%
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
Molecular Formula C6H5F3O2
MDL Number MFCD08060542
CAS Min % 96.5
Assay Percent Range 97%
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material [5-(Trifluoromethyl)-2-furyl]methanol
CAS 65865-28-3

Specifications

Chemical Name or Material [5-(Trifluoromethyl)-2-furyl]methanol
CAS 65865-28-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H5F3O2
Formula Weight 166.1
MDL Number MFCD08060542
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.