missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Trifluoromethyl)-2-furaldehyde, ≥97%, Maybridge

$528.03 - $528.03

Chemical Identifiers

CAS 17515-80-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H3F3O2
Formula Weight 164.08
MDL Number MFCD00525585
Chemical Name or Material 5-(Trifluoromethyl)-2-furaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC49804CB SDS Fisher Scientific Company
CC49804CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC49804DA SDS Fisher Scientific Company
CC49804DA
Amber Glass Bottle 1g Each for $528.03
Specifications Description & Specifications

Chemical Identifiers

17515-80-9
100.0
C6H3F3O2
MFCD00525585
97%
96.5
97%
164.08
5-(Trifluoromethyl)-2-furaldehyde

Specifications

250mg
96.5
Amber Glass Bottle
C6H3F3O2
MFCD00525585
97%
17515-80-9
100.0
97%
164.08
5-(Trifluoromethyl)-2-furaldehyde

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning