missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Trifluoromethyl-1,3-phenylenediamine, TCI America™

Manufacturer:  TCI America T32395G

 View more versions of this product

Catalog No. T32395G



Specifications

Specifications

5-Trifluoromethyl-1,3-phenylenediamine
89°C
368-53-6
MFCD00014776
KZSXRDLXTFEHJM-UHFFFAOYSA-N
5-(trifluoromethyl)benzene-1,3-diamine
67781
Crystal-Powder at 20°C
Undesignated
5g
C7H7F3N2
3,5-diaminobenzotrifluoride, 5-trifluoromethyl benzene-1,3-diamine, 5-trifluoromethyl-1,3-phenylenediamine, 5-trifluoromethyl-1,3-benzenediamine, 3,5-benzotrifluorodiamine, unii-z89rd529kk, 5-trifluoromethyl-benzene-1,3-diamine, 1,3-benzeneamine, 5-trifluoromethyl, 1,3-benzenediamine, 5-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-3,5-diamine
C1=C(C=C(C=C1N)N)C(F)(F)F
176.142
176.14
≥98.0% (GC,T)

Chemical Identifiers

368-53-6
176.142
KZSXRDLXTFEHJM-UHFFFAOYSA-N
67781
C1=C(C=C(C=C1N)N)C(F)(F)F
C7H7F3N2
MFCD00014776
3,5-diaminobenzotrifluoride, 5-trifluoromethyl benzene-1,3-diamine, 5-trifluoromethyl-1,3-phenylenediamine, 5-trifluoromethyl-1,3-benzenediamine, 3,5-benzotrifluorodiamine, unii-z89rd529kk, 5-trifluoromethyl-benzene-1,3-diamine, 1,3-benzeneamine, 5-trifluoromethyl, 1,3-benzenediamine, 5-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-3,5-diamine
5-(trifluoromethyl)benzene-1,3-diamine
Safety and Handling

Safety and Handling

RTECSNumber : CZ1606000

TSCA : No

Documents