Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Trifluoromethoxy)salicylic acid, 98%

$278.61 - $706.53

Chemical Identifiers

Chemical Name or Material 5-(Trifluoromethoxy)salicylic acid
CAS 129644-57-1
Molecular Formula C8H5F3O4
Formula Weight 222.12
MDL Number MFCD01091003
Synonym 2-Hydroxy-5-(trifluoromethoxy)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2609903

 
alfa aesar™
H2609903
1g Each for $706.53

AAH26099MD

 
alfa aesar™
H26099MD
250mg Each for $278.61
Description & Specifications

Specifications

Formula Weight 222.12
Chemical Name or Material 5-(Trifluoromethoxy)salicylic acid
Assay 98%
Molecular Formula C8H5F3O4
MDL Number MFCD01091003
CAS 129644-57-1
Synonym 2-Hydroxy-5-(trifluoromethoxy)benzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 131° to 133°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.