Please login to your online account to display your discounted pricing

5-(Trifluoromethoxy)isatin, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC425970010

 View more versions of this product

Catalog No. AC425970010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 231.13
Quantity 1g
CAS Max % 100.0
CAS Min % 96.0
Chemical Name or Material 5-(Trifluoromethoxy)isatin
CAS 169037-23-4
Molecular Formula C9H4F3NO3
Assay Percent Range 97%
MDL Number MFCD00192524
Packaging Glass bottle

Specifications

Chemical Name or Material 5-(Trifluoromethoxy)isatin
CAS 169037-23-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H4F3NO3
Formula Weight 231.13
MDL Number MFCD00192524